Learn More
Ace Glass, Inc. Combo mantle, 3 position, low temperature, fits 650mL round bottom flasks, max diameter 4.25 inches, 120v 200 watts per position, order code (-61) or (-71) power supply separately
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ace Glass, Inc. 12061113
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Combo mantle, 3 position, low temperature, fits 650mL round bottom flasks, max diameter 4.25 inches, 120v 200 watts per position, order code (-61) or (-71) power supply separately