Learn More
3M™ 6000 Series Probed Reusable Respirator Assembly
Probed reusable respirator assembly 6000Q series are used for quantitative fit purposes only.
$52.27
Specifications
Product Type Quantitative Fit Test Assembly
Certifications/Compliance EN 140:1998, NIOSH 42 CFR Part 84
For Use With (Application) Conduct Quantitative Fit Tests on Respirator 6000 Series
For Use With (Equipment) Conduct Quantitative Fit Tests on Respirator 6000 Series
Includes Probed facepiece and P100 filter
View More Specs
Products 2
Catalog Number Mfr. No. Model Size Price Quantity & Availability  
Catalog Number Mfr. No. Model Size Price Quantity & Availability  
17-987-3A
View Documents
3M™
7000126030
6200 Q Medium
Each for $52.27
Case of 4 EA for $209.10
Add to Cart
17-987-3B
View Documents
3M™
7000126031
6300 Q Large
Each for $52.27
Case of 4 EA for $209.10
Add to Cart
Description
Description
Combines features of both reusable and disposable respirators to provide effective protection. 3M™ Half Face Mask Respirator 6000 Series are economical, low-maintenance, simple-to-handle, and extremely lightweight.
  • Probed reusable respirator assembly 6000 Series used for quantitative fit testing
  • Probed facepiece allows for numerical measurement of the amount of leakage into the respirator and includes a probed facepiece and P100 filters
Compliance
NIOSH approved under 42 CFR 84 standard for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters; in Canada, CSA standard Z94.4 and applicable local government requirements must be met for approval.
Specifications
Specifications
Quantitative Fit Test Assembly
Conduct Quantitative Fit Tests on Respirator 6000 Series
Probed facepiece and P100 filter
EN 140:1998, NIOSH 42 CFR Part 84
Conduct Quantitative Fit Tests on Respirator 6000 Series
Thermoplastic Elastomer