Learn More
CPC Scientific 5TAMRA-Lys-Lys-Leu-Asn-Arg-Thr-Leu-Ser(PO3H2)-Val-Ala-OH (trifluoroacetate salt) 1MG
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CPC Scientific PROK008B
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
ONE-LETTER SEQUENCE: 5TAMRA-KKLNRTL-Ser(PO3H2)-VAMOLECULAR FORMULA: C74H113N18O21P1MOLECULAR WEIGHT:1621.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases