Learn More
Electron Microscopy Sciences 5 Microscope Slide Container Model 2
Supplier: Electron Microscopy Sciences 7154908
Holds up to 5 slides 3x1"in size . These mailers are rigid, strong and durable with a convenient flip-top, snap-on lid. When it stands in its upright position, it can be used as a staining container, with a minimal amount of solution being used.