Learn More
US Plastic Corporation 5 GALLON NATURAL MOLDED POLYET
SDP
Supplier:  US Plastic Corporation 15749
Encompass
5 Gallon Natural Molded Polyethylene Tamco® Tank with Lid & 1/2" FNPT Fitting - 18" L x 9" W x 10" Hgt.
Catalog No. NC2205275
Explore available promotions