Learn More
McKesson General Medical Burn Dressing Water-Jel First Responder 4 X 4 Inch Square Sterile, 60/cs
Supplier: McKesson General Medical 1071032
Pre-saturated with an odor-free, crystal clear gel. The clear gel allows for clear visualization and assessment of burn injuries at any time. Water-Jel First Responder burn dressings contain a patented technology thickener, stabilizer and suspension which is: Benzene-free, self wetting, non-adherent, clear.
This item is not returnable.
View return policy