Learn More
Scott Safety™ SKA-PAK™ Supplied Air Respirators
Offers emergency respiratory protection for variety of applications.
$196.21 - $2140.00
Specifications
Product Type | Supplied Air Respirator System |
---|---|
For Use With (Application) | Maintenance work, hazardous materials handling, turn-arounds and confi ned space operations, chemical, food processing, petrochemical, oil & gas, ship building and public utilities |
Pressure | 2216 psig |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
19-122-013
|
Scott Safety™
SAR222010211101 |
AV-2000, Dupont™ Kevlar™ aramid fiber (CS) facepiece |
Each
for $196.21
Save 91% Reg : |
|
|||||
19-122-011
|
Scott Safety™
SAR222010211401 |
AV-2000 (CS) Poly facepiece |
Each for $2,140.00
|
|
|||||
19-122-010
|
Scott Safety™
7012623426 |
No Facepiece | N/A | N/A | |||||
Please contact your local Fisher Scientific sales representative for pricing | |||||||||
Description
- Designed for use in hazardous atmospheres
- Combination Type C positive pressure respirator with escape cylinder for entry into/escape from hazardous, confined space or IDLH situations
- Includes E-Z Flo pressure demand regulator with first-breath activation and low exhalation
- 5-, 10-, or 15-minute rated cylinder egress options for selection versatility based on task at hand
- Hip-mounted cylinder with durable, secure cylinder band retention system
- Available in two harness styles:
- Polypropylene - chemically resistant and lightweight
- Kevlar - Forhigh heat environments or when there is potential for sparking
- Available with Hansen, Schrader, Cejn, Hansen HK plug or Hansen HK socket fittings
- Can be used with AV-2000, AV-3000 or AV-3000 with SureSeal facepieces
Applications:
Maintenance work, hazardous materials handling, turn-arounds and confined space operations.Used in chemical, food processing, petrochemical, oil & gas, ship building and public utilities.
Specifications
Supplied Air Respirator System | |
Maintenance work, hazardous materials handling, turn-arounds and confi ned space operations, chemical, food processing, petrochemical, oil & gas, ship building and public utilities | |
2216 psig |