Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Disposable Protective Overshoe Covers
Click to view available options
Blue
Description
Reliable, disposable boot cover designed to help protect footwear against light liquid splashes and hazardous dusts. Made from a microporous laminate material with an anti-static treatment.
- Covers are designed with ties and elastic at the calf to help ensure a secure fit
- Distinctive unisex design
- 3M Catalog# 402
Specifications
Specifications
Size | Universal |
Material | Polypropylene |
Color | Blue |
Disposable | Single-use |
Closure Type | Elastic Ankle |
Model | 402 |
For Use With (Application) | Assembly, Cleaning, Machine Operations |
Resistance | Dry Particulates, Fluid |
Product Type | Boot Covers |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction