Learn More
World Precision Instrument NanoFil Needles-NF35BL-2
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: World Precision Instrument NF35BL2
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The replaceable needles used with the NanoFil are available with either blunt or beveled tips. The blunt tip is used for injection into soft tissue and when a uniform solution distribution is needed. The beveled style is used for applications that involve the penetration of a tough tissue.