Learn More
Enzo Life Sciences 3-O-Methyl-Sphingomyelin (1 mg)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Enzo Life Sciences BMLSL2250001
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
3-O-Methyl-Sphingomyelin is a novel selective inhibitor of neutral sphingomyelinase, (Ki=37 µM). It is cell-permeable and does not inhibit acid sphingomyelinase. Purity: ≥98% (TLC). Solubility: Soluble in ethanol (>25 mg/ml) or DMSO (20 mg/ml@50°C). Long Term Storage: -20°C.