Learn More
Consolidated Plastics 3.5-7 GALLON PEEL OFF LID-WHIT
SDP
Supplier:  Consolidated Plastics 33450
Encompass_Preferred
Consolidated Plastics brings you their reusable easy peel bucket lid. This peel off pail lid is compatible with 3.5, 5, 6 and 7 gallon buckets. Made from FDA approved HDPE (high density polyethylene) resin. 70 mil heavy duty construction.
Catalog No. NC2027776
Explore available promotions