Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Impacto™ 3/4-Finger Anti-Fatigue Thermo Gloves
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Description
- Natural pain relief for prevention of injuries during work or play
- Designed to allow full ventilation during repetitive tasks, for extended periods of time
- Light but firm therapeutic compression counteracts tissue swelling to provide temporary relief from repetitive strain injuries (RSI)
- Promotes optimal muscle function at 1°C higher than the body’s normal resting temperature
- Open-finger design maximizes tactile feedback
- Ideal for repetitive jobs such as sewing, or for wearing inside other gloves when handling frozen products
Specifications
Specifications
Color | Black |
Material | Nylon |
Size | 2X-Large |
Finger Style | Three Quarter |
For Use With (Application) | Temporary Pain Relief from Repetative Strain Injuries (RSI) |
Glove Cuff Style | Hook and Loop |
Series | TS199 |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction