Learn More
Chemglass Life Sciences Replacement 25.4mm Stirrer Shaft, Reactor Size: 30L, Fits Support Frame: CG-1968-X-12, Overall Length: 40.9"
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG209733
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Replacement 25.4mm Stirrer Shaft, Reactor Size: 30L, Fits Support Frame: CG-1968-X-12, Overall Length: 40.9". Precision ground and polished stirrer shaft for use with Chemglass process reactors. Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a 3/8 wide flat. REPLACEMENT STIRRER SHAFTS ARE FOR USE WITH MECHANICAL SEAL STIR BEARING ON LISTED SUPPORT FRAME.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.