Learn More
Electron Microscopy Sciences Critical Point Dryer 24 Specimen Holder
Supplier: Electron Microscopy Sciences 7019424
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Machined from solid steel and brass/nickel plated. Incorporates a stainless steel mesh top and bottom to allow for fluid exchange. Comes with two sections; each section holds 12 specimens in 12 cavities cavity measures 4.5mm(D) x 6mm(H)..