Learn More
AGA Marvel 24-IN GENERAL PURPOSE REFRIGERATOR
Supplier: AGA Marvel MS24RAS4RW
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
1 diameter probe port in rear cabinet wall (standard) 3 full-depth perforated metal shelves, cantilever mounted on rear wall of interior, adjustable in 1.5 increments Flat interior door liner maximizes depth of shelves Box Style door for easy mounting of inventory management system hardware Front vented through toe space grill for true built- in installations Extra-long, jacketed commercial power cord with hospital grade plug to meet stringent hospital and clinical use standards CSA Approved and warrantied for commercial use Standard door lock for secure storage needs Discrete top edge mounted door handle Cam-assisted door close hinge system to reduce door ajar events Bright energy efficient white LED interior lighting for easy content viewing 4 leveling legs for up to 1 height adjustment Durable white powder coat paint finish
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.