Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxytoluene, 98%, Thermo Scientific Chemicals

Catalog No. AAB2260614
Click to view available options
Quantity:
25 g
100 g

2,6-Dimethoxytoluene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethoxytoluene is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5673-07-4

Specifications

Melting Point 37°C to 41°C
Boiling Point 222°C
Flash Point 96°C (204°F)
Odor Aromatic
Molecular Formula C4H7ClMg
MDL Number MFCD00674003
Quantity 25 g
Beilstein 2045327
Synonym methallylmagnesium chloride, 2-methylallylmagnesium chloride, magnesium, chloro 2-methyl-2-propenyl, methallyl magnesium chloride, methallyl-magnesium chloride, 2-methylallyl magnesium chloride, bjvfgwybolmuem-uhfffaoysa-m, 2-methylallylmagnesium chloride solution, 2-methyl-2-propenyl magnesium chloride, 2-methyl-2-propen-1-yl magnesium chloride
Solubility Information Insoluble in water.
InChI Key BJVFGWYBOLMUEM-UHFFFAOYSA-M
SMILES CC(=C)C[Mg]Cl
IUPAC Name magnesium;2-methanidylprop-1-ene;chloride
Molecular Weight (g/mol) 114.86
PubChem CID 5175691
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 2,6-Dimethoxytoluene
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 227-131-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.