Learn More
Electron Microscopy Sciences 2.5% Glutaraldehyde in Sorensons Buffer, 7.2 pH 10 ML
Supplier:  Electron Microscopy Sciences 15980
Encompass_Preferred
Premixed, prebuffered fixatives, are pH adjusted for immediate laboratory use. They are freshly prepared upon receipt of order to ensure maximum shelf life. The fixatives are filled in 20 ml glass vials (10 ml/vial). This allows for the immediate fixation of your specimens without costly and time consuming transfer associated with ampoule contained fixatives. In Sorensons Buffer.
pH: 7.2
Catalog No. 50-259-46