Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethylphenylmagnesium bromide, 1M in 2-MeTHF, Thermo Scientific Chemicals

Catalog No. AAH54282AE
Click to view available options
Quantity:
100 mL

 

2,4,6-Trimethylphenylmagnesium bromide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylphenylmagnesium bromide is used as an organic chemical synthesis intermediate.

Solubility
It reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2633-66-1
Molecular Formula C9H11BrMg
Molecular Weight (g/mol) 223.40
MDL Number MFCD00009663
InChI Key VZMGGVXGYWKQPB-UHFFFAOYSA-M
Synonym 2-mesitylmagnesium bromide, mesitylmagnesium bromide, 2,4,6-trimethylphenylmagnesium bromide, 2-mesitylmagnesium bromide solution, mesmgbr, yxvsitsudrgill-uhfffaoysa-m, magnesium,bromo 2,4,6-trimethylphenyl, 2-mesitylmagnesium bromide solution, 1.0 m in thf, 2,4,6-trimethylphenylmagnesium bromide, 1m in 2-methf, 2-mesitylmagnesium bromide solution, 1.0 m in diethyl ether, grignard reagent
PubChem CID 5098599
IUPAC Name magnesium;1,3,5-trimethylbenzene-6-ide;bromide
SMILES CC1=CC([Mg]Br)=CC(C)=C1C

Specifications

Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Solubility Information It reacts with water.
Formula Weight 223.4
Concentration or Composition (by Analyte or Components) 1M in 2-MeTHF
Chemical Name or Material 2,4,6-Trimethylphenylmagnesium bromide
Hazard Category H225-H302-H314-H335
Hazard Statement GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.