Learn More
Waters Corp Polypropylene 12 x 32 mm Snap Cap Vial, with Cap and PTFE/Silicone Septum, 700 µL Volume, 100/pk
Supplier: Waters Corp 186005223
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Clean polypropylene-molded vial, preferred in applications where there is a concern that compound or molecule may stick to a polar glass surface. This vial is a cost effective alternative to glass.