Learn More
Hans Rudolph Inc Series 1140 SmartLab™ Instrumentation System with InSight™software
SDP
Supplier:  Hans Rudolph Inc 263001
Encompass
Request Bulk Order
The SmartLabTM with Insight™ software is a flexible respiratory measurement and data collection system for use by researchers. It is a modular system that has many configuration options. This base unit can integrate with up to four different sensor modules as listed below. It also has inputs for a Nonin xPod oximeter, Masimo IRMA CO2 sensors both in-line and sidestream sampling and a temperature sensor. An O2 sensor sampling system can be integrated. There are voltage input modules for ±5 & ±10 VCD as well as a 4 channel ±10volt analog output system. Radio Frequency (RF) communications is available if needed. A laptop computer can be supplied with all software installed.
Catalog No. NC2120619