Learn More
Bioassay Systems QuantiQuik™ Glucose Quick Test Strips. Direct determination of glucose concentrations in food and beverage samples as well as biological (e.g. urine, blood, etc) samples.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Bioassay Systems QQGLUC10
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For rapid, direct determination of glucose concentrations in food and beverage samples as well as biological (e.g. urine, blood, etc.) samples. Key Features: Fast and sensitive. Use of 20 or 100 µL sample. Semi-quantitative measurement between 0-720 mg/L (undiluted) Glucose. Convenient. No expensive lab equipment needed. Sample trea™ent and assay can be performed in under 15 minutes. Method: Visual. Samples: Wine, beer, juice, etc. Species: All. Procedure: Assay takes approximately 15 min. Kit size: 10 tests.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.