Learn More
Chemglass Life Sciences 100mL Peptide Synthesis Vessel, Solid Phase, Medium Frit, GL 25 Thread
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG186104
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
100mL Peptide Synthesis Vessel, Solid Phase, Medium Frit, GL 25 Thread. Solid phase peptide synthesis vessel having a medium or coarse porosity fritted glass resin support. Top of the vessel has a GL thread and is supplied complete with a PTFE lined PBT screw cap. Side port and lower PTFE stopcock have a 2mm bore with the arms being 8mm I.D.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.