Learn More
Bioassay Systems EnzyChrom™ Phospholipase D Assay Kit. For quantitative determination of phospholipase D activity and evaluation of drug effects on phospholipase D metabolism.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Bioassay Systems EPPD100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For quantitative determination of phospholipase D activity and evaluation of drug effects on phospholipase D metabolism. Key Features: Sensitive. Use 10 µL samples. Detection range: colorimetric assay 0.06 - 10 U/L, fluorimetric assay 0.04 - 1 U/L. Simple and High-throughput: the assay involves addition of a single working reagent and can be readily adapted to high-throughput assays for drug screening. Method: OD570nm, or FL530/585nm. Samples: Biological. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 0.04 U/L.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.