Learn More
Bioassay Systems QuantiChrom™ Indole Assay Kit. Indole determination in biological samples.
Supplier: Bioassay Systems DIND100
For quantitative determination of indole and evaluation of drug effects on its metabolism. Key Features: Fast and sensitive. Use of 100µL sample. Linear detection range from 3 to 100 µM indole in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, and reading the absorbance immediately. Method: OD565nm. Samples: Bacterial growth media and other biological samples. Species: All. Procedure: Assay takes 5 min. Kit size: 100 tests. Detection limit: 3 µM.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.