Learn More
Bioassay Systems EnzyFluo™ Isocitrate Assay Kit. For quantitative determination of isocitrate and evaluation of drug effects on its metabolism.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Bioassay Systems EFIC100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For quantitative fluorimetric determination of isocitrate and evaluation of drug effects on its metabolism. Key Features: Fast and sensitive. Linear detection range (20 µL sample): 0.6 to 500 µM for 10 min reaction. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: FL530/585 nm. Samples: Food, beverage, biological samples (e.g. cell lysate, tissue homogenate, serum, etc.). Species: All. Procedure: Assay takes 10 min. Kit size: 100 tests. Detection limit: 0.6 µM.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.