Learn More
Bioassay Systems QuantiFluo™ Alkaline Phosphatase Assay Kit. For quantitative determination of alkaline phosphatase (ALP) activity using stable 4-methylumbelliferyl phosphate substrate.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Bioassay Systems QFAP100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For quantitative determination of alkaline phosphatase (ALP) activity using stable 4-methylumbelliferyl phosphate substrate. Key Features: High sensitivity and wide linear range. Use 10 µL sample. Detection limit of 0.02 U/L (20 min reaction). Homogeneous and simple procedure. Simple "mix-and-measure" procedure allows reliable quantitation of ALP activity within 20 minutes. Robust and amenable to HTS. All reagents are compatible with high-throughput liquid handling instruments. Method: FL360/450nm. Samples: Serum, plasma etc. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: 0.02 U/L.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.