Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ 10% Neutral Buffered Formalin in Seal-Rite Containers

Use these containers for the transportation and short-term storage of patient specimens.

Supplier:  Epredia™ LC0020

 View more versions of this product

Catalog No. 22-045-400



Description

Description

Features:
  • Seal-Rite containers are engineered to maintain seal at 95KPA – the industry standard for proper sealing and transportation via air shipment
  • Double-click feature provides audible confirmation that the lid has been aligned, tightened properly and sealed
  • Arrow and lock icons align to visually signify the containers have been properly sealed
  • Embossed mL measuring scale identifies volume of liquid in the container
  • Knurled cap and base of the jar for better gripping
  • Safety. Security. Peace of Mind – Avoid the loss of precious tissue samples and exposure of staff to chemicals caused by leaking containers
A properly closed Epredia Seal-Rite container is leak-proof.
The containers have been tested using industry-standard International Safe Transit Association (ISTA) performance validation. Internal tests were performed as well with 0% leakage rate.
Specifications

Specifications

Tray; 784/cs; Cs. dimensions 25-5/16 x 15-7/16 x 13-1/8 in; Cs. weight 38 lbs
Formalin
20mL
Safety and Handling

Safety and Handling

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit