Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Cartridges for Multidrop™ Pico 1 & Pico 8 Digital Dispensers
The Dispensehead cartidges allow dispensing of one to eight fluids per cassette, from 11 picoliters to 10 microliters of small molecules and biomolecules in DMSO or aqueous solution
Supplier: Thermo Scientific™ LTR0002
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Description
Great choice for dispensing picoliter to microliter volumes for one to eight different types of fluid across an entire plate or just part of a plate.
- Cost Saving through assay miniaturization
- Improved efficiency and sample integrity
- Powerful, flexible, and easy to use software
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Digital Dispenser Cartridge | |
1-ch 200μL Cartridges for Multidrop Pico 1 | |
11 pL to 20 μL, 1 nL to 200 μL | |
Mulitdrop Pico 1 Digital Dispenser | |
12, 24, 48, 96, 384, 1536 well plate, Plate height: 6-47 mm | |
PicoIT 1 | |
8.2 x 6.5 X 2.2 cm | |
100 to 240V 50/60Hz | |
50/60 Hz |
Not Autoclavable | |
1 | |
Drug combination studies, Biochemical assays, Cell-based assays, DMPK, Secondary screening, Synthetic chemistry, assay development, qPCR, Enzyme profiling, Antibody therapies, SAR, and Spotting | |
USB | |
≤ 8% | |
11 pL to 10 μL in seconds | |
0.225 | |
100/240 V |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction