Thermo Scientific Vari-Mix and Speci-Mix Test Tube Rockers
Offer a choice of fixed - or adjustable-angle models.
Features:
- All models hold 10 to 30mm diameter tubes and are ideal for blood collection tubes; Choose from fixed or adjustable speeds
- Reversible pad provides one side (with lip to prevent sliding) for capped tubes 110mm (4.33in.) long; other side for longer tubes
- Autoclavable white silicone rubber pad enhances viewing
- Can be used in a noncondensing incubator or coldroom with ambient operating conditions of 4° to 40°C (39.2° to 104°F) and 20 to 80% RH noncondensing
Thermo Scientific MaxQ 8000 Shakers
This video showcases the Thermo Scientific MaxQ 8000 Shakers.
Ease-Of-Use
Video Highlights of the Thermo Scientific MaxQ 8000 Shakers ease-of-use.
Easy Cleaning and Maintenance
Thermo Scientific MaxQ 8000 Easy Cleaning and Maintenance Video.
Ergonomic
Thermo Scientific MaxQ 8000 shakers - features an ergonomic fold down door and handle.
Flexible
The Thermo Scientific Max Q 8000 shaker plantforms featuer exceptional flexability.
HEPA Protection
Demonstration of the easily accessible HEPA Filter.
Intuitive Controls
MaxQ 8000 easy to program intuitive controls.
Sample Visibility
Large window and internal light allows for sample viewing.
Service Made Easy
Electronic Components and HEPA filter are accessed from the front of the shaker.
Thermo Scientific CO2 Resistant Digital Compact Bottle/Tube Roller
Maximize valuable bench space with our compact, adjustable-speed unit, capable of rolling multiple tube and bottle sizes.
Features
- Adjustable speed accommodates a variety of applications
- Digital speed control from 1 -80 RPM
- Small footprint ideal for use in a CO2 incubator
- Anti-corrosive and wear-resistant construction enables use in humid and CO2 environments
- Adjustable rollers accommodate a variety of tube/bottle sizes
- Stacking kit allows you to stack units two high, saving valuable bench space
- Optional remote control facilitates operating unit while in incubator and add timer function
- Includes pack of O-rings for tubes. O-rings ensure consistent smooth rolling when using tubes
Applications
Cell Culture | Hybridization Assays | Liquid Mixing
Thermo Scientific Digital Tube Revolver
Increase your lab's efficiency with our variable-speed tube revolver. Its small footprint, flexible design and simple display help increase flexibility and effectiveness.
Features
- Interchangeable rotisseries accommodate different tube sizes for maximum flexibility. Each paddle can be adjusted from 0-90 degrees
- Achieve more effective mixing with "oscillating" mode. Oscillation will occur at every 40 degrees in the rotation cycle
- Small footprint
- Plug-and-play simplicity
- Maintenance-free motor, quiet operation
Standard Unit Includes:
- 90-691-36 — 10mL/15mL paddle, Quantity 2
- 90-691-37 — 5mL/7mL paddle, Quantity 2
- 90-691-38 — 1.5mL/2mL paddle, Quantity 2
- 90-691-40 — 50mL paddle, Quantity 1
Applications
Molecular Biology | Clinical Biochemistry
Thermo Scientific Compact Digital Waving Rotator
Achieve maximum sample mixing efficiency with the adjustable orbital waving digital-display rotator.
Features:
- PID control ensures consistent and smooth rocking motion
- Digital Speed Control: 8 to100rpm
- Easy to adjust tilt angle: 0° to 13°
- Compact design ideal for placing in incubators (non-CO2), chambers and refrigerators (up to 60°C)
- Timer range: continuous or 1 minute to 99 hours 59 minutes
- Includes dimpled mat (tray, silicone pad, two securing bands)
- When no angle is used, the Waving Rotator can be used as a slow speed orbital shaker
Applications
Designed for time-dependent and low-dependent sensitive applications such as ELISA, protein synthesis, staining/de-staining gels and antibody staining.
Thermo Scientific Cel-Gro Tissue Culture Rotator
Available in a single or dual drum configuration.
Features
- Single-drum as an adjustable rotating angle and speed, while the dual-drum has a fixed rotating angle and speed
- Rotator operates in standard incubators for cultivation under controlled temperature conditions
- Powder-coated stainless-steel outer body construction is durable and corrosion-resistant
- Drums can be lifted off the rotor without interrupting rotator operation
- Adjusting screw on the base varies elevation angle
- Adjustable 5° rotating angle above horizontal to 90°
- Adjustable speed control from 12 to 70rpm
- Drum required with purchase, sizes vary based on tube size requirements:
- 142 x 17mm tube diameter
- 76 x 26mm tube diameter
- 60 x 30mm tube diameter
Thermo Scientific LP Vortex Mixer
Choose to continuously mix or pulse samples with our low-profile vortex mixer.
Features:
- Low-profile design with small footprint
- Run in continuous or pulsing mode
- Speed control: 0 to 300rpm
- Automatic overload shut off
- Wide variety of optional accessories to accommodate different vessel types
Applications
Any application that requires intense vortex mixing in a tube, plate or glass vessel.
Thermo Scientific MaxQ Shakers Brochure
The Thermo Scientific MaxQ Shaker product selector guide will help you build the optimal shaker for your lab and application.
Thermo Scientific Rockers, Rotators and Shakers Brochure
The Thermo Scientific Rockers, Rotators and Shakers brochure includes a list of products along with ordering information and product specifications.