Honeywell North Powered Air Purifying Respirators and Accessories
Engineered to be ergonomic, efficient, and effortless, the Honeywell North™ Primair™ 700 (PA700) Series Powered Air Purifying Respirator (PAPR) keeps workers comfortable and productive.
The PA700 Series PAPR was designed with these features and benefits in mind:
Convenience
- Easy to assemble – components lock into place
- Easy-to-operate controls – even when wearing gloves
- Easy to clean – smooth surfaces take just minutes to wipe down
Comfort
- Reduces the need for breaks or adjustments to equipment
- On-demand airflow control keeps workers cool
- Additional accessories available to enhance comfort
Confidence
- Integrated, streamlined design is reliable and durable
- Components that lock into place make workers feel safer
- Assigned protection factor (APF) of 1000 with bibbed hood; APF of 25 with loose-fitting head cover