ASTM Testing Standards
ASTM International is recognized globally for developing and publishing voluntary standards. More than 12,000 ASTM standards are currently used worldwide to improve product quality, enhance health and safety, strengthen market access and trade, and build consumer confidence.
ASTM Standards
The ASTM defines a standard as “a document that has been developed and established within the consensus process of the Society and that meets the approval requirements of ASTM procedures and regulations.”
The ASTM publishes six types of standards:
- Test method
- Specification
- Classification
- Practice
- Guide
- Terminology
ASTM test methods are definitive procedures that produce a result. They usually include a detailed description of a procedure for determining a property or constituent of a material, an assembly of materials, or a product. These details include the apparatus, test specimen, procedure, and calculations needed for satisfactory results.
ASTM test methods represent a consensus about the best currently available procedure and are supported by experience and data from cooperative tests.
Resources
Trace Elemental Analysis
Numerous techniques can be used to determine trace or ultra-trace level impurities in substances or materials, an analysis that is especially useful to manufacturers of foods and beverages, polymers and plastics, adhesives and coatings, metals, pharmaceuticals, electronics, and many other consumer and industrial products.
To help eliminate false readings due to contamination, choose specially purified chemicals and reagents for your sample preparation and analytical methods.