Food and Beverage Quality Control Testing
In today’s food and beverage production, testing of critical quality control parameters is more important than ever before.
Measuring quality throughout the manufacturing process is essential to ensure raw materials arrive as expected before use, intermediate components achieve production requirements, and finished products meet safety and quality specifications for consumer sale and approval. In the pursuit of safer, higher-quality, and longer-lasting foods and beverages, quality control regulations are tightening, all while consumer cost expectations are decreasing. Food and beverage manufacturers are challenged to achieve higher output targets with increased efficiency and reduced testing errors to improve profits.
Leveraging over 50 years of experience with electrochemistry testing, Thermo Fisher Scientific has partnered with food and beverage producers throughout the world to help make their complex quality measurements routine, reliable, and accurate.
The range of Thermo Scientific Orion pH, Ion, Conductivity, and Dissolved Oxygen Meters and Thermo Scientific ROSS Electrodes allows quality control managers to reduce the operational cost of critical measurements, maintain laboratory proficiency, and return significant process efficiency improvements for the business through intuitive instrument operation for fast sample measurement and high-performance electrodes designed to improve accuracy, reproducibility, and stability in food and beverage samples.
Thermo Scientific Orion Star T900 Series Titrators are designed to make performing titrations easier, more reliable, and more reproducible than manual titrations. Our automated titrators significantly reduce the time analysts spend standing in front of the titration equipment, remove the need for color indicators and analyst-interpretation of endpoints, and automatically perform result calculations and data logging. Each titrator stores up to 10 customizable methods so routine procedures can be repeated without additional setup or pre-work, and methods can be shared among titrators using a USB flash drive.
Since most food and beverage samples require testing for acidity, our automated Orion Star T910 pH Titrator is paired with our high-performance ROSS pH Electrode and recommended for a wide variety of food and beverage samples.
Content provided by: