Bioclean-D Drop-Down-Garment
World's First True Aseptic Gowning Technique for Disposable Coveralls
Constructed from antistatic, low-linting durable CleanTough fabric, these coveralls feature an innovative drop-down design and quick-release tab system that enable fast, 100% touch-free donning, with no risk of cross contamination, resulting in greater productivity.