Thermo Scientific Industrial Microbiology Products
Supporting Safe Manufacturing of Food and Pharmaceutical Products
Find Thermo Scientific™ industrial microbiology testing essentials that support the manufacturing of safe pharmaceuticals and food and beverage products. Count on these complete, market-leading solutions to detect microbes in your environment, raw materials, intermediaries, and finished products.
Thermo Scientific microbiology products for pharma and biopharma applications are designed to address the unique needs of these industries. They deliver a range of validated solutions for the pharmaceutical microbiology laboratory, supporting the production and quality assurance of sterile pharmaceuticals, vaccines, and biotechnology products.
Environmental Monitoring
Meet your environmental monitoring needs with sealed, sterile, and secure Thermo Scientific Triple Wrap Tryptone Soya AgarPlates. Pharmacopoeia-compliant, they are available in both 60 mm contact and 85 mm settling plates.
- Isolator compatible: Three protective wraps, including a VHP™ impervious layer, enhance media quality and protection
- Achieve full traceability: Includes 2D (GS1-matrix) barcoding for sampling accuracy
- Improve efficiency: Reduce time spent on incoming goods controls with large-volume single lots
- Enhance sustainability: Reduce waste with Styrofoam-free shipper packaging and practical storage temperatures (2 to 25°C) with no refrigeration required
Triple Wrap Sterile Pack Tryptone Soya Agar with VHP Indicator and 2D Barcode
Sterility Testing
Find high-performance media with an extensive selection of formats and a broad range of volumes, from 20 mL to 1,000 mL, which support the growth of a wide variety of bacteria, yeasts, and molds.
- Quality components: Manufactured with premium raw materials and filled in ISO-5 cleanroom suites
- Extensive range: Complete USP/EP formulated media for sterility workflows; single-lot orders available
Microbial Limits Testing and Quality Control
Browse a comprehensive range of dehydrated and ready-to-use media and quality control organisms to support testing from raw materials to finished goods.
- Globally harmonized: Aligned to USP/EP/JP formulations and QC releases per compendial methods
- Media for all specified organisms: Bile-tolerant, Gram-negative bacteria, aerobic and anaerobic bacteria, molds, and fungi
- Complete and robust: Quality control microorganisms for growth promotion testing
Aseptic Process Simulations
Enable positive outcomes by using a sterile growth medium as your placebo. Reduce burden and stay in control of aseptic manufacturing processes using compliant, validated Thermo Scientific Cold Filterable Media.
- Minimize preparation: Easy and user-friendly preparation–cold filterable media means no heating required
- Reduce production downtime: Dehydrated culture media has excellent solubility, reducing the risk of blockages
- Save time: Reduce time on incoming goods control with large-volume single lots
Explore microbiology solutions that bring together the best in food quality and safety testing. They include market-leading molecular instrumentation and PCR technology for foodborne pathogen and quality indicator detection as well as traditional culture media and biochemical or immunological tests.
Enrichment Media and Sample Preparation
Bring simplicity and accuracy to your laboratory with flexible Thermo Scientific™ Oxoid™ enrichment media formats combined with sample dilutors and homogenizers.
- Work more efficiently: Prepare media in a matter of minutes
- Eliminate steps: No need for weighing and autoclaving
- Protect your environment: Save storage space and reduce waste
- Reinvent the process: Dilute samples quickly and accurately at the touch of a button
Rapid Detection with PCR
Detect common food pathogens rapidly and accurately with the Thermo Scientific™ SureTect™ Real-Time PCR System. It offers an easy-to-use workflow and provides results in 8 to 26 hours—depending on the target and test matrix—which is 3 to 6 days faster than traditional culture-based methods. SureTect is ISO and AOAC certified/validated for a wide range of food matrices.
- Easy handling: Ready-to-use reagents and ergonomically designed tools
- Stay on track: Color-coded plates and reagents
- Simplify your workflow: Analyze for multiple pathogens simultaneously
- Smart instrumentation: Automate your lysis, test setup, and result interpretation
Rapid Detection with Chromogenic Media
Simplify detection, enumeration, and identification of Salmonella, Listeria, and Cronobacter with Thermo Scientific™ Precis™ Methods that enable rapid, culture-based workflows.
- Minimize handling: Single overnight enrichment
- Reduce costs: Single chromogenic plate medium
- Improve efficiency: Simple rapid confirmation and identification options
- Maintain consistency: Harmonized workflow for all Precis methods
Confirmation, Identification, and Quality Control
Find accurate, simple, rapid, and convenient solutions to suit your immunological, immunological, or molecular identification needs. Save time and reduce the risk of infection and contamination during QC testing with Thermo Scientific™ Culti-Loops™ direct-streak inoculating loops.
These ready-to-use, gel-preserved microorganisms do not require rehydration or organism handling and give you confidence with complete traceability of ATCC™* License Derivative and NCTC cultures.
*The ATCC Licensed Derivative™ Emblem, the ATCC Licensed Derivative™ word mark, and the ATCC catalog marks are trademarks of ATCC. Remel Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures. The identity, purity, and authenticity of the Licensed Products are exclusively the responsibility of Remel Inc. and not ATCC.
Triple Wrap Plates Introduction Video
Find out how Thermo Scientific Triple Wrap Plates for Environmental Monitoring provide greater assurance for your sterile products.
Quanti-Cult Kits Demonstration Video
Learn how Thermo Scientific Quanti-Cult Plus kits can provide certainty in the quality control of your pharmaceutical microbiology processes.
SureTect Introduction Video
Learn how to simplify your food pathogen testing workflow and get faster results.
Listeria Precis Introduction Video
Discover the next generation in culture media-based testing and simplify your workflow.
More Resources
Pharma Quality Control Organisms Brochure
Find out how to gain confidence in your growth promotion with Thermo Scientific Quanti-Cult Plus Quantitative Quality Control Microorganisms kits.
- Pharma Quality Control Organisms Brochure (784.9 KB)
Triple Wrap Plates Brochure
Discover pharmacopoeia-compliant Thermo Scientific Triple Wrap Plates that are designed to support your most sterile environments.
- Triple Wrap Plates Brochure (657.7 KB)
Pharma Bottled Media Brochure
Explore Thermo Scientific Bottled Media varieties designed to support the growth of a wide range of bacteria, yeasts, and molds.
- Pharma Bottled Media Brochure (1.5 MB)
SureTect PCR Range Brochure
Learn how to simplify your food pathogen testing with the Thermo Scientific SureTect PCR system.
- SureTect PCR Range Brochure (4.6 MB)
Listeria Precis Methods Brochure
See how Thermo Scientific Listeria Precis Methods reinvent the standard for culture-based Listeria testing.
- Listeria Precis Methods Brochure (1.1 MB)