Thermo Scientific Biological Safety Cabinets
The Difference Is the Design
With Thermo Scientific™ biological safety cabinets, you never have to think about user and sample protection. Designed with Thermo Scientific™ SmartFlow™ technology, the cabinets include dual-DC motors to automatically balance the cabinet inflow and downflow air velocities in real time—even as the filters load with contaminants. Plus, the Digital Airflow Verification (DAVe) alarm signals any out-of-spec conditions for added assurance. Combined with the reliability, ergonomic features, and energy efficiency of Thermo Scientific brand products, the ideal choice is also the one you can trust completely. And not just on day one.
Innovations in Containment and Efficiency
A biological safety cabinet's primary function is to provide cleanliness and containment. Features available on Thermo Scientific biological safety cabinets include:
- SmartFlow dual-motor design to automate airflow compensation and balance
- Digital Airflow Verification (DAVe) to alert users to excessive flow restriction
- Night Set-Back mode that reduces airflow to 30% when window is closed
- A 75% reduction in energy use and a longer motor and filter life than traditional AC motor cabinets
- Reduced noise level enhances user focus
- 10° sloped front for exceptional ergonomics
- Spacious work area with comfortable armrest
- Increased light intensity for brighter work environment
- SmartClean front window design for easy cleaning
- Intuitive control panel for at-a-glance monitoring
- SmartPort organizes tubing and cables with convenient side access
- Programmable UV lights extends bulb life and saves energy
Product Details
- The built-in WiFi system* and touchscreen are ready for cloud-based capabilities like remote equipment monitoring, secure data storage, collaboration tools, and more
- Glass side walls and LED lighting make for a brighter workspace with better visibility
- SmartClean Plus motorized, hinged front window design enables easycleaning
- Dual DC motors power the SmartFlow Plus airflow compensation system while delivering better energy efficiency and quieter operation
- The crossbeam UV light system features an exclusive self-adjusting exposure capability to enable reproducible germicidal effectiveness with every use
- The SmartPort allows you to easily pass tubing and wiring through the sidewall to accommodate application needs
- Work surfaces can be customized to suit your application and work style
Product Details- The built-in WiFi system* and touchscreen are ready for cloud-based capabilities like remote equipment monitoring, secure data storage, collaboration tools, and more
View More Thermo Scientific Biological Safety Cabinets
*Availability of connectivity-enabled models depends on region. Please contact your sales representative to learn more.
Thermo Scientific Herasafe 2030i Class II A2 Biological Safety Cabinet
The Herasafe 2030i is equipped with “smart” self-monitoring safety features, an intuitive graphic touchscreen, and built-in connectivity to preserve essential performance data.
Learn More About the Thermo Scientific Herasafe 2030i Class II A2 Biological Safety Cabinet
Thermo Scientific 1300 Class II A2 Biological Safety Cabinet
This biological safety cabinet helps protect users and keep them comfortable with a proprietary airflow design, ergonomic features, and energy efficiency for operational cost savings.
Learn More About the Thermo Scientific 1300 Class II A2 Biological Safety Cabinet
Thermo Scientific Biological Safety Cabinets Airflow Alarms Flyer
Why are airflow alarms an important consideration for Biological Safety Cabinets (BSCs)?
Thermo Scientific Biological Safety Cabinets DC Motor Flyer
Why should you select a Class II Biological Safety Cabinet (BSC) with a DC motor over one with an AC motor?
Thermo Scientific Biological Safety Cabinets Dual Blower Flyer
Why is a dual blower system better than a single blower system?
Thermo Scientific Biological Safety Cabinets Filter Life Flyer
How do I extend the filter life of my biological safety cabinet (BSC)?
More Resources
Thermo Scientific Biological Safety Cabinets Operating Costs Data Sheet
Managing groups of biological safety cabinets (BSCs) requires an optimization strategy that addresses both performance and cost.