Workplace Safety Products
Explore equipment, supplies, and other laboratory safety products and hospital safety products to help you comply with best practices for taking care of patients with respiratory illnesses. Choose from a wide variety of personal protective equipment, including products for social distancing, hand sanitation, disinfecting shared areas, temperature checks, and more.
Featured Products
Limit virus contact and transfer with fundamental supplies, including basic face and hand protection.
General-Purpose Face Masks
Disposable face masks are designed for single or limited use for particulate filtration and fluid resistance. Universally sized, face masks may have ties or soft, elastic ear loops and 2- or 3-ply construction. They are not airtight, do not require fit testing, and are not intended for medical use.
Procedure and Surgical Masks
Surgical masks are tested to meet levels of barrier protection as specified in ASTM F2100-19. Procedure masks are FDA approved medical devices for use in healthcare procedures. They are not airtight and do not require fit testing.
Fisherbrand Face Shield
Lightweight and comfortable, Fisherbrand™ Face Shields provide protection from splashing liquids, blood, and other bodily fluids and can be used in laboratories, medical settings, dentist offices, and veterinary practice.
N95 Respirators
An N95 mask or respirator filters out at least 95% of airborne particles as defined by the U.S. National Institute for Occupational Safety and Health (NIOSH, 42 CFR Part 84). These essential hospital safety products are functionally equivalent to FFP2 respirators (European Union) and KN95 respirators (China) with slightly different certification criteria.
Read MoreSurgical N95 Respirator
Surgical respirators, also called Surgical N95, medical, or healthcare respirators, are N95 masks that meet both FDA and NIOSH requirements for healthcare use. During COVID-19 pandemic shortages, the Families First Coronavirus Response Act changed the liability and certification requirements to allow the use of regular N95 respirators in healthcare settings.
Read MorePowered Air-Purifying Respirators (PAPRs)
Surgical respirators, also called Surgical N95, medical, or healthcare respirators, are N95 masks that meet both FDA and NIOSH requirements for healthcare use. During COVID-19 pandemic shortages, the Families First Coronavirus Response Act changed the liability and certification requirements to allow the use of regular N95 respirators in healthcare settings.
Read MoreGloves and Industrial Gloves
Protect hands in the clinical lab or at the point of care with durable gloves that offer comfort, tactile sensitivity, dexterity, and puncture resistance.
Safety Signs
Notify patients and colleagues that wearing face masks or gloves is required in your hospital or clinical laboratory.
Biohazardous Waste Disposal
Safely contain and control biohazardous materials in your lab or facility.
Encourage good personal hygiene with easily accessible hand sanitation stations.
Hand Sanitizer Dispensers
Social Distancing Aids
Provide the visual cues and barriers needed to maintain appropriate distancing in high-traffic, meeting, and other shared spaces.
Effectively sanitize shared areas with essential cleaning tools and supplies.
Disinfectant Cleaners
General-Purpose Floor Cleaning Products
Squeegees
Contamination Control
Use temperature checks to monitor potential health issues along with signage and tents to help manage safety and screening procedures.