Multiplex Testing Solutions for COVID-19 and Influenza
Find a variety of tests for detecting influenza and SARS-CoV-2 using a single test. Learn more.
In today’s dynamic healthcare environment, the need for accurate and efficient diagnostics is crucial, especially during surges of influenza and COVID-19. The Fisher Healthcare channel offers a robust portfolio of point-of-care assays based on molecular and PCR technology, as well as reader systems, to help healthcare providers detect these illnesses using cost-effective multiplex testing kits and solutions.
Explore a curated selection that provides dependable detection of respiratory pathogens, supports streamlined clinical workflows, and offers greater cost control and convenience.
 Abbott ID NOW COVID-19 2.0 Test KitRoche Diagnostics cobas liat SARS-CoV-2 + Influenza A/B Multiplex AssayQuidel Sofia 2 Flu+SARS Antigen Fluorescent Immunoassay (FIA)BD Veritor System for Rapid Detection of SARS-CoV-2 & Flu A+BVisby Medical Respiratory Health Test
 Abbott ID NOW™ COVID-19 2.0 Test KitRoche Diagnostics™ cobas™ liat™ SARS-CoV-2 + Influenza A/B Multiplex AssayQuidel™ Sofia™ 2 Flu+SARS Antigen Fluorescent Immunoassay (FIA)BD Veritor™ System for Rapid Detection of SARS-CoV-2 & Flu A+BVisby Medical™ Respiratory Health Test
ShopBuy ID NOW COVID-19 Test KitsBuy cobas liat SARS-CoV-2 + Influenza A/B AssaysBuy Quidel Sofia 2 Flu+SARS Antigen FIAsBuy BD Veritor Systems for SARS-CoV-2 & Flu A+BBuy Visby Medical Respiratory Health Tests
Catalog Number23-046-65523-312-20523-044-155B25608823-111-203
DetectsSARS-CoV-2 only or SARS-CoV-2 and influenza A and B with the same patient swab.*SARS-CoV-2 and influenza A and BSARS-CoV-2 and influenza A and BSARS-CoV-2 and influenza A and BSARS-CoV-2 and influenza A and B
MethodNucleic Acid AmplificationRT-PCRAdvanced Immunofluorescence-Based Lateral FlowLateral Flow ImmunoassayRT-PCR
Time to Result5 Minutes (Positive)
13 Minutes (Negative)
20 Minutes15 Minutes15 Minutes30 Minutes
Required InstrumentAbbott ID NOW InstrumentRoche Diagnostics cobas liat SystemQuidel Sofia 2 Fluorescent Immunoassay AnalyzerBD Veritor Plus SystemInstrument-free
*ID NOW Influenza A & B 2 test kits sold separately. ID NOW software update to version 7.1 required for sequential workflow capability.