Clinical Chemistry and Immunoassay Analyzers
To meet your needs for more automation, broader test menus, and increased efficiencies, the Fisher Healthcare channel has partnered with trusted clinical analyzer manufacturers and suppliers. This enables us to stay on the cutting edge of technology and serve as informed consultants to aid in your decision making. Get access to multiple clinical chemistry and immunoassay instruments to meet your day-to-day and future diagnostic testing needs.
See the current portfolio of clinical chemistry and immunoassay analyzers available from:
Complete the form and a Fisher Healthcare account manager will contact you to help you choose an analyzer that best suits your laboratory’s needs.
By submitting your data, you agree to our Privacy Policy and will receive emails based on your preferences.
© Thermo Fisher Scientific, Inc. All Rights Reserved.
Vitros XT3400 Chemistry Systems
The QuidelOrtho™ Vitros™ XT3400 Chemistry System helps you double your output with 3D imaging that rapidly detects and deletes outliers.
- Performs up to 1,130 chemistry tests per hour
- Requires only 74.8 µL
- No water supply access required
- Direct ion-selective electrode (ISE) assays
Vitros 4600 Chemistry Systems
The comprehensive test menu of the QuidelOrtho™ Vitros™ 4600 Chemistry System helps your lab support the diverse diagnostic needs of today’s clinicians.
- Assay ranges may be configured for up to seven demographic attributes.
- Dry slide technology
- Vitros Microslide filters out proteins and lipids to reduce interfering substances
- Vitros MicroTips eliminate carryover
Vitros 3600 Immunodiagnostic Systems
The QuidelOrtho™ Vitros™ 3600 Immunodiagnostic System has enhanced chemiluminescence detection and a comprehensive test menu that covers all major disease states.
- Throughput of 189 immunoassay tests per hour
- 150 reagent positions
- Vitros MicroSensor detects any endogenous interferences
- Vitros INTELLICHECK monitors, verifies, and documents diagnostic checks throughout the testing process
Vitros ECiQ Immunodiagnostic Systems
The QuidelOrtho™ Vitros™ ECiQ Immunodiagnostic System offers a comprehensive menu of routine and specialty immunodiagnostic testing, a throughput of up to 90 tests per hour, and a compact footprint.
- Vitros MicroWell enhanced Chemiluminescence Detection
- 150 reagent positions
- Vitros INTELLICHECK monitors, verifies, and documents diagnostic checks throughout the process
Vitros XT7600 Integrated Systems
The QuidelOrtho™ Vitros™ XT7600 Integrated System offers a full chemistry and immunoassay menu, with automation scaling options and valuable informatics features.
- Sample size: 2.7 µL
- Processes a test every three seconds
- Up to 1,320 tests per hour with XT MicroSlides
- Test menu of over 160 assays
Vitros 5600 Integrated Systems
The QuidelOrtho™ Vitros™ 5600 Integrated System is designed to run over 900 clinical chemistry and immunoassay tests per hour.
- Consolidate testing on a single analyzer
- First-pass yield of 96.5%
- Perform tests independently and in parallel — no transport or aliquoting
- No water supply access required
- Test menu of 120 assays
IMMULITE Immunoassay Analyzers
The continuous random-access Siemens Healthineers™ IMMULITE™ 2000 XPi immunoassay analyzer can process up to 200 tests per hour. Its ability to perform specialty and allergy testing on a single platform enhances productivity for medium- to high-volume laboratories. Customize the automated startup for daily routine maintenance and schedule your quality control.
Yumizen C1200 AL Analyzers
The Horiba™ Yumizen™ C1200 AL Analyzers perform up to 1,200 tests per hour with ISEs. It has random continuous access and a dedicated STAT position.
- Refrigerated reagent (90) and calibrator/control (61) positions
- Liquid level sensing, clot detection
- Checks for hemolysis, icterus, and lipemia
- Optional autoloader (sold separately)
Pentra C400 Analyzers
The Horiba™ Pentra™ C400 Analyzer incorporates spectrophotometry, colorimetry, turbidimetry, and potentiometry.
- 52 reagent positions (44 refrigerated, 8 ambient)
- Maximum throughput: 420 tests per hour
- Optional ISE module
- Integrated work and validation station
AU480 Chemistry Analyzers
The compact, reliable, and cost-effective Beckman Coulter™ AU480 Chemistry Analyzer offers a versatile and efficient clinical chemistry solution. With a throughput of up to 400 photometric tests (up to 800 with ISEs) per hour, the AU480 can be your primary analyzer in low- to mid-volume hospitals and laboratories or a dedicated specialty or STAT analyzer in larger labs.
- More than 120 different assays
- STAT wheel for priority testing
- Sample pre-dilution and auto-repeat for over-range results
- Ready-to-use concentrated liquid reagents
- Low consumable use
DxC 700 AU Chemistry Analyzers
Get the right results at the right time. Suited for mid- to high-volume labs, the Beckman Coulter™ Unicel™ DxC 700 AU combines the popular features of the DxC and AU clinical chemistry analyzers.
AU5800 Chemistry Analyzers
Give your high- and ultra-high volume core laboratory a performance boost with AU5800 Series Clinical Chemistry Analyzers. It’s capable of processing up to 2,000 tests per hour with a full assay menu, including TDM and DAT. Scalable up to four modules.
- Completely replace sample and reagent probes, mixer bars, ISE, and syringes in less than 60 seconds
- Easy routine maintenance
- Modular design lets you Increase testing capacity as needed
- Rapid TAT for STATs and auto-repeat of out-of-range result
Access 2 Immunoassay Systems
Maximize your laboratory's productivity with the Beckman Coulter™ Access™ 2 Immunoassay System, a benchtop analyzer with a space-saving design and a menu of more than 50 tests. Updated features include desktop sharing, reagent pack barcode reading, and ergonomic design.
- 100 test per hour throughput
- Load 60 samples at one time
- Random access and automatic reflex testing
- Proven chemiluminescent and magnetic particle separation technology
UniCel Dxl 600 Access Immunoassay Systems
The Beckman Coulter™ UniCel™ DxI™ 600 Access™ Immunoassay System meets the needs of laboratories running 40,000 to 80,000 tests per year with its 200 tests per hour throughput. Streamlined maintenance procedures and “load and go” sample loading saves time for other important tasks.
- Standardized test menus, assay protocols, instrument processes, and reference ranges
- Broad test menu
- Can be integrated with automation and IT systems
UniCel Dxl 800 Access Immunoassay Systems
Shorten assay time-to-results in your high-volume lab with the Beckman Coulter™ UniCel™ DxI™ 800 Access™ Immunoassay System. A 400 tests per hour throughput, streamlined maintenance procedures, and “load and go” sample loading saves time for other important tasks.
- Standardized test menus, assay protocols, instrument processes, and reference ranges
- Broad test menu
- Can be integrated with automation and IT systems
Phadia 250 Laboratory Systems
The Thermo Scientific™ Phadia™ 250 Laboratory System delivers 400 to 2,000 results per week, is fully automated, and designed for medium-sized laboratories. It provides allergy and autoimmunity testing solutions using state-of-the-art technology.
Phadia 1000 Laboratory Systems
The Thermo Scientific™ Phadia™ 1000 Laboratory System meets the needs for allergy testing in large, cost-conscious laboratories, producing 2,000 to 10,000 results per week.
Phadia 2500 Laboratory Systems
Provide optimal allergy and autoimmunity testing solutions using state-of-the-art technology with the Thermo Scientific™ Phadia™ 2500 Laboratory System. It manages very high throughputs of 10,000 to 25,000 results per week.