Learn More
Fisherbrand™ Standard Serum Filters
Used when the filtered serum can be immediately poured off for analysis.
$77.00
Specifications
Product Type | Serum Filter |
---|---|
For Use With (Application) | Used when filtered Serum can be immediately poured off for analysis |
For Use With (Equipment) | Glass collection tubes |
Catalog Number | Diameter (English) Outside | Diameter (Metric) Outside | Length (English) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Diameter (English) Outside | Diameter (Metric) Outside | Length (English) | Length (Metric) | Price | Quantity & Availability | |||
02-681-53
|
0.4 in. | 10.25mm | 4 in. | 100mm |
Pack of 100 for $77.00
Case of 40 PK for $2,403.00
|
|
|||
02-681-50
|
0.5 in. | 13mm | 4 in. | 100mm |
Pack of 100 for $77.00
Case of 40 PK for $2,403.00
|
|
|||
02-681-52
|
0.6 in. | 16mm | 6 in. | 152mm |
Pack of 100 for $77.00
Case of 40 PK for $2,403.00
|
|
|||
02-681-51
|
0.6 in. | 16mm | 4 in. | 100mm |
Pack of 100 for $77.00
Case of 40 PK for $2,406.00
|
|
|||
02-681-58
|
0.4 in. | 10.25mm | 2 in. | 20mm | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | |||
Description
Used when the filtered serum can be immediately poured off for analysis.
Disposable devices for filtering fibrin from the serum portion of centrifuged blood samples contained in standard 10.25, 13 or 16mm I.D. collection tubes.
- Standard model is similar to Fisherbrand™ IB™ model, but without the one-way valve
- Use only with glass collection tubes
- Technique reduces second centrifugation, tube labeling and sample preparation time
- Allows direct sampling into automatic laboratory equipment
- Eliminates fibrin, particulate matter and equipment downtime
Specifications
Serum Filter | |
Used when filtered Serum can be immediately poured off for analysis | |
Glass collection tubes |