Learn More
VistaLab™ Technologies MLA™ Silver Brand Precision Pipetters
Made of precision engineered aluminum alloy and stainless steel with limited Lifetime Warranty (covering parts). Individually serialized with Calibration Certificate included.
$384.00 - $390.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Color | Silver |
Material | Aluminum Alloy, Stainless Steel |
Model | MLA Precision |
Number of Channels | 1 |
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
22-147-608
|
VistaLab™ Technologies
1101 |
5 μL |
Each for $386.00
|
|
|||||
22-147-613
|
VistaLab™ Technologies
1102 |
15 μL |
Each for $386.00
|
|
|||||
22-147-614
|
VistaLab™ Technologies
1103 |
30 μL |
Each for $384.00
|
|
|||||
22-147-615
|
VistaLab™ Technologies
1104 |
40 μL |
Each for $390.00
|
|
|||||
22-147-616
|
VistaLab™ Technologies
1106 |
60 μL |
Each for $384.00
|
|
|||||
22-147-617
|
VistaLab™ Technologies
1107 |
70 μL |
Each for $384.00
|
|
|||||
22-147-618
|
VistaLab™ Technologies
1108 |
75 μL |
Each for $386.00
|
|
|||||
22-147-619
|
VistaLab™ Technologies
1109 |
80 μL |
Each for $386.00
|
|
|||||
22-147-620
|
VistaLab™ Technologies
1110 |
90 μL |
Each for $384.00
|
|
|||||
22-147-621
|
VistaLab™ Technologies
1111 |
120 μL |
Each for $390.00
|
|
|||||
22-147-622
|
VistaLab™ Technologies
1113 |
400 μL |
Each for $386.00
|
|
|||||
22-147-609
|
VistaLab™ Technologies
1115 |
600 μL |
Each for $386.00
|
|
|||||
22-147-610
|
VistaLab™ Technologies
1117 |
800 μL |
Each for $386.00
|
|
|||||
22-147-611
|
VistaLab™ Technologies
1118 |
900 μL |
Each for $390.00
|
|
|||||
22-147-612
|
VistaLab™ Technologies
1119 |
450 μL |
Each for $386.00
|
|
|||||
22-147-678
|
VistaLab™ Technologies
1505 |
700 μL |
Each for $390.00
|
|
|||||
Description
- “To deliver” capability ensures volume accuracy over a wide range of viscosities
- Ideal for repetitive delivery of reagents when high precision and accuracy are required
- Capacity engraved on bonnet
Specifications
Not Autoclavable | |
Aluminum Alloy, Stainless Steel | |
1 | |
Precision Pipetter | |
1 Ea. | |
9 in. | |
1.5 in. |
Silver | |
MLA Precision | |
Non-sterile | |
Nozzle Insert, Calibration Key | |
Limited Lifetime | |
2 in. |