Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caplugs™ Evergreen Scientific BMC/Hitachi™ 717, 736, 737 Analyzer Sample Cups

For use with BMC/Hitachi Analyzer models 717, 736, and 737

Supplier:  Caplugs™ 1271212B10

Catalog No. 05-555-1


Explore available promotions
Add to Cart
This item is not returnable. View return policy

Description

Description

  • Capacity: 2.0 to 2.5mL
  • Material: Polystyrene
  • Color: Blue

Specifications

Specifications

Sample Cup
2 to 2.5 mL
BMC/Hitachi analyzer Models 717, 736, 737
Polystyrene
Blue
Automated chemistry analyzers
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.