Refrigerators
Frequently used in research, clinical, and industrial settings, laboratory refrigerators are manufactured for both general and special purposes.
Read More About RefrigeratorsTop Categories
Convenient, Daily Storage
Laboratory Refrigerators
General purpose units designed for daily use in research, clinical, and industrial labs. Ideal for storing non-critical products such as reagents, media, and proteins.
Cool Columns with Confidence
Chromatography Refrigerators
These specialized units provide temperature-controlled environments for protecting chromatography columns during separation. They may include alarms for temperature change, door ajar, or power failure.
Safely Store Volatile Materials
Flammable and Explosion Proof Refrigerators
Include features to prevent fires and explosions when storing volatile materials like solvents and intermediates. Electrical components are separate from refrigerator contents to prevent sparks.