Methylene Chloride
solvents-methylene-chloride-banner-ch2cl2-d-20-396-2074
What Is Methylene Chloride?
Methylene chloride, also called dichloromethane (DCM), is a volatile, polar, organochloride compound that’s miscible with many organic solvents.
A colorless liquid at room temperature, methylene chloride has a sweet, penetrating odor. It’s a member of the halomethanes or chloromethanes group where one or more hydrogens has been replaced by a halogen (chlorine).
Though non-combustible and non-flammable, methylene chloride can emit toxic chloride fumes when exposed to high temperatures. The vapors are narcotic in high concentrations.
Dichloromethane is formally rated as a probable carcinogen by IARC 2A and is a potentially toxic compound. It has been detected in feces, saliva, and other biofluids.
Synonyms: Dichloromethane, methylene dichloride, methylene bichloride, DCM
View All Methylene Chloride Products
solvents-methylene-chloride-image-ch2cl2-20-396-2074
Physical and Chemical Properties
PropertyValue  
Property
Molecular Weight
Value
84.93 g/mol
Property
Density
Value
1.3255 g/mL at 20ºC
Property
Boiling Point
Value
40ºC
Property
Melting Point
Value
-95.1oC
Property
Flash Point
Value
5.5oC
Property
Viscosity
Value
0.413 cP at 25ºC
Property
Dielectric Constant
Value
8.93 at 25oC
Property
Dipole Moment
Value
1.6
Property
UV Cutoff
Value
233 nm
Property
Refractive Index
Value
1.4244 at 20oC
Property
Vapor Pressure
Value
435 mm Hg at 25oC
Property
Water Solubility
Value
13 mg/mL at 25°C
Property
Heat of Combustion
Value
28.82 x 106 J/kg at 25oC
Property
Surface Tension
Value
28.2 dyne/cm at 20oC
Uses
Methylene chloride is commonly used as a solvent for organic reactions and extractions, and in drug and pesticide manufacturing. It’s also used as a refrigerant and paint stripper and to remove caffeine from coffee and tea.
Designations and Grades
PropertyValue  
Property
CAS Number
Value
75-09-2
Property
MDL Number
Value
MFCD00000881
Property
RIDAD/UN Number
Value
1593
Property
Beilstein Registry
Value
1730800
Property
EG/EC Number
Value
200-838-9
Property
NSC Number
Value
406122
Property
ICSC Number
Value
0058
Property
RTECS Number
Value
PA8050000
Grades
ACS/ACS Reagent/Certified ACS
Anhydrous/Dry/Extra Dry/Low Water
Biochemical
Environmental
FCC
GC/Capillary GC
GC/Pesticide Residue
GC-MS
HPLC
Laboratory/Technical
NF
Pesticide
Spectrophotometry