Hexane
solvents-hexane-banner-c6h14-d-20-396-2073
What Is Hexane?
Hexane, also called hexanes, is a mixture of straight-chain alkanes with six carbon atoms. It consists of approximately 60 percent n-hexane, varying amounts of the isomers 2-methylpentane and 3-methylpentane, and smaller amounts of non-isomer alkanes. Hexane is colorless and odorless when pure and has a boiling point of roughly 69°C (156°F).
This solvent cannot be easily deprotonated and is therefore used in labs for reactions involving very strong bases, like preparing organolithiums. Hexane can also extract oil and grease from water and soil. In chromatography, hexane is used as a non-polar solvent, but its alkanes may interfere with analyses if the sample becomes contaminated.
Hexane is obtained primarily by crude oil refinement and its composition depends on the source of the oil and the refining process.
View All Hexane Products
solvents-hexane-image-c6h14-20-396-2073
Physical and Chemical Properties
PropertyValue  
Property
Molecular Weight
Value
86.18 g/mol
Property
Density
Value
0.6606 g/mL 25°C
Property
Boiling Point
Value
68.73°C
Property
Melting Point
Value
-95.3°C
Property
Flash Point
Value
-22°C
Property
Viscosity
Value
0.300 cP at 25ºC
Property
Dielectric Constant
Value
1.890 at 20°C
Property
UV Cutoff
Value
210 nm
Property
Refractive Index
Value
1.3727 at 25°C
Property
Vapor Pressure
Value
13 mm Hg at 20°C
Property
Water Solubility
Value
0.0095 mg/mL at 25°C
Property
Heat of Combustion
Value
4163.2 kJ/mol
Property
Heat of Vaporization
Value
1.56 kJ/mol at 25°C
Property
Surface Tension
Value
17.89 mN/m at 25°C
How Is Hexane Used?
Hexane is used in textile manufacturing to make glues and adhesives for shoes, leather products, and roofing. It’s also used for cleansing and degreasing and to extract cooking oils, like canola or soy, from seeds.
Designations and Grades
PropertyValue  
Property
CAS Number
Value
110-54-3
Property
MDL Number
Value
MFCD00009520
Property
RIDAD/UN Number
Value
1208
Property
Beilstein Registry
Value
1730733
Property
EG/EC Number
Value
203-777-6
Property
NSC Number
Value
8472
Property
ICSC Number
Value
0279
Property
RTECS Number
Value
MN9275000
Grades
ACS/ACS Reagent/Certified ACS
Biochemical
Dry/Extra Dry/Molecular Sieve
Electronic
Environmental
GC/Pesticide Residue/HR-GC
HPLC/HPLC Gradient/LC-MS
Laboratory/Technical
Pesticide
Spectrophotometry