Learn More
ZeptoMetrix NATtrol™ Adenovirus Type 5
SDP
Qualitative molecular control
Supplier:  ZeptoMetrix NATADV5ST
Catalog No. 22-157-767
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Formulated with purified, intact virus particles that have been chemically modified to render them non-infectious and refrigerator stable
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
Specifications
Specifications
ISO 13485
Adenovirus Type 5 Control (qualitative)
1 mL
2°C to 8°C
Evaluate the performance of Nucleic Acid Tests for determination of the presence of Adenovirus Type 5 DNA and also for validation of clinical assays, development of diagnostic Tests and training of laboratory personnel