Learn More
ZeptoMetrix NATtrol™ Adenovirus Type 41
SDP
Qualitative molecular control
Supplier:  ZeptoMetrix NATADV41ST
Catalog No. 22-157-765
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Formulated with purified, intact virus particles that have been chemically modified to render them non-infectious and refrigerator stable
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
Specifications
Specifications
ISO 13485
Adenovirus Type 41 Control (qualitative)
1 mL
2°C to 8°C
Evaluate the performance of Nucleic Acid Tests for determination of the presence of Adenovirus Type 41 DNA and also for validation of clinical assays, development of diagnostic Tests and training of laboratory personnel