Learn More
Enzo Life Sciences Yellow Fever NS1, (recombinant) (100µg)
Supplier: Enzo Life Sciences ENZPRT1090100
Alternative name: YFV NS1. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In DPBS pH 7.4. Source: Produced in HEK 293 cells. Yellow Fever Virus NS1 protein from strain 17D. Long Term Storage: -80°C. Applications: ELISA Vaccine research, POC diagnostic, LINE assay, Antibody capture.