Learn More
Agilent Technologies XL1-BLUE MR SUPERCOMP CELLS
Supplier: Agilent Technologies 200229
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Cells; XL1-Blue MR Supercompetent; Stratagene; For cloning w/o F episome, greater-than-equal-to 1 x 109 transformants/microgram; Deficient in all known E. coli K12 restriction systems; Increase efficiency of introducing eukaryotic DNA