Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ Women's Navy Flame Resistant Flex Knit Button Down Shirt
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- The best of two hard-working worlds, this lightweight FR Flex Knit combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt.
- Lightweight stretch fabric and side seam gussets flex with your body for added comfort and range of movement.
- Two chest pockets with button flaps allows convenient storage.
- Easy-to-spot CAT 2/2112 label on the left sleeve leave no question that you're protected on the job site.
- Certifications Level 1: NFPA 2112, NFPA 70E, ASTM F1506
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Arc Flash, Flash Fire
- Number of Pockets: 2
- Primary Closure: Button
- Wash Care: Industrial Laundry - Light Soil
- Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey
- Blend: Body: 93% Cotton / 7% Spandex Elastane
- Collar: Banded Collar
- Pocket: Two chest pockets with flaps and button closure
- Features: Banded collar, Full underarm gusset for added mobility, Lightweight, stretch fabric, Two chest pockets with flap and sewn in pencil stall
- Protection: ATPV Arc Rating: 8.3
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Feminine Fit
- Gender: Female
- Brand: Bulwark™
Specifications
Size | 2X-Large |
Gender | Women's |
Material | Cotton, Spandex |
Color | Navy |
Neck Style | Collar |
Sleeve Style | Long Sleeve |
Construction | Knit |
Closure Type | Button |
Disposable | Reusable |
Hazard Risk Category | Category 2 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction