Learn More
Electron Microscopy Sciences White Porcelain Plate
Supplier:  Electron Microscopy Sciences 7156206
Encompass_Preferred
12 cavities on a white porcelain plate. Used for staining and color reactions. They measure: 4 5/8"(L) x 3 1/2"(W) (118x90 mm). Cavity depth: 1/4" (6.4 mm).
Catalog No. 50-931-197